Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Chlorothieno[3.2.c]pyridine 95%

Bottle, 1 g
Catalog # APOSOR30584-1G
Irritant
Supplier:   Apollo Scientific
4-Chlorothieno[3.2.c]pyridine 95%
Catalog # APOSOR30584-1G
Supplier:  Apollo Scientific
CAS Number:  27685-94-5
4-Chlorothieno[3.2.c]pyridine 95%

Bottle, 1 g

CAS Number:  27685-94-5
Price
102,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4-Chlorothieno[3,2-c]pyridine
  • CAS Number:27685-94-5
  • Formula:C₇H₄ClNS
  • MDL Number:MFCD01861830
  • Molecular Weight:169.63 g/mol
  • Melting Pt:97…100 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications