
3-Bromothieno[3,2-c]pyridine
Catalog # APOSOR30582-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:214.08 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:28783-18-8
Formula:C7H4BrNS
MDL Number:MFCD11869740