Order Entry
Export
ContactUsLinkComponent
3-Iodo-1,5-naphthyridine
3-Iodo-1,5-naphthyridine
Catalog # APOSOR305571-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Iodo-1,5-naphthyridine
Catalog # APOSOR305571-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :256.05 g/mol
Storage Temperature:Ambient
CAS Number:1228666-28-1
Formula:C₈H₅IN₂
MDL Number:MFCD16628231