4,4'-Dihydroxyazobenzene
Supplier: Apollo Scientific
About this item
Synonyms: 4,4-Dihydroxyazobenzene, 4,4'-Azodiphenol
- CAS Number:2050-16-0
- Formula:C₁₂H₁₀N₂O₂
- MDL Number:MFCD00045778
- Molecular Weight:214.22 g/mol
- Melting Pt:215 °C
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: 4,4-Dihydroxyazobenzene, 4,4'-Azodiphenol
- CAS Number:2050-16-0
- Formula:C₁₂H₁₀N₂O₂
- MDL Number:MFCD00045778
- Molecular Weight:214.22 g/mol
- Melting Pt:215 °C
- Storage Temperature:Ambient
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New Product
More
Specifications
Cat. No.APOSOR305565-250MGNew Product - Specifications:
More
Specifications
Cat. No.APOSOR305565-1G - Specifications:New Product
More
Specifications
Cat. No.APOSOR305565-5GNew Product - Specifications:New Product
More
Specifications
Cat. No.APOSOR305565-25GNew Product - Specifications:New Product
More
Specifications
Cat. No.APOSOR305565-100GNew Product



