Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Phenylene diisocyanate
1,3-Phenylene diisocyanate 98%
1,3-Phenylene diisocyanate 98%
1,3-Phenylene diisocyanate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
1,3-Phenylene diisocyanate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
123-61-5
:
Formula:
C₈H₄N₂O₂
MDL Number:
MFCD00002018
Storage Temperature:
Ambient