1H-Pyrrolo[3,2-c]pyridine-4-carboxylic acid
Catalog # APOSOR305346-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:162.15 g/mol
Storage Temperature:Ambient
CAS Number:1040682-84-5
Formula:C₈H₆N₂O₂
MDL Number:MFCD09965904