6-Nitroimidazo[1,2-a]pyridine
Catalog # APOSOR305165-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:163.1300 g/mol
Storage Temperature:Ambient
CAS Number:25045-82-3
Formula:C₇H₅N₃O₂
MDL Number:MFCD05863333