Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Quinoxaline-2,3-dione
Quinoxaline-2,3-dione 98%
Quinoxaline-2,3-dione 98%
Quinoxaline-2,3-dione 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Quinoxaline-2,3-dione 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
15804-19-0
:
Formula:
C₈H₆N₂O₂
MDL Number:
MFCD00006723
:
162.15 g/mol
Storage Temperature:
Ambient