Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-1,4-naphthoquinone
2-Chloro-1,4-naphthoquinone 97%
2-Chloro-1,4-naphthoquinone 97%
2-Chloro-1,4-naphthoquinone 97%
APOSOR30485-100G
:
Apollo Scientific
:
2-Chloro-1,4-naphthoquinone 97%
APOSOR30485-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
192.60 g/mol
:
Ambient
:
1010-60-2
:
C₁₀H₅ClO₂
:
MFCD00029187