Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(+)-Methyl 3-hydroxybutyrate
(S)-(+)-Methyl-3-hydroxybutyrate
(S)-(+)-Methyl-3-hydroxybutyrate
(S)-(+)-Methyl-3-hydroxybutyrate
Supplier:
Apollo Scientific
Certificates
Expand image
(S)-(+)-Methyl-3-hydroxybutyrate
Supplier:
Apollo Scientific
About this item
CAS Number:
53562-86-0
:
Formula:
C₅H₁₀O₃
MDL Number:
MFCD00064461
:
118.13 g/mol
Storage Temperature:
Ambient