Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Cyclohexyl-1H-imidazo[4,5-b]pyridine
1-Cyclohexyl-1H-imidazo[4,5-b]pyridine
1-Cyclohexyl-1H-imidazo[4,5-b]pyridine
1-Cyclohexyl-1H-imidazo[4,5-b]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
1-Cyclohexyl-1H-imidazo[4,5-b]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
1365094-43-4
:
Formula:
C₁₂H₁₅N₃
MDL Number:
MFCD22126109
:
201.27 g/mol
Melting Pt:
161…164 °C
Storage Temperature:
Ambient