Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4,6-dichloro-5-nitronicotinate
Methyl-4,6-dichloro-5-nitronicotinate 95%
Methyl-4,6-dichloro-5-nitronicotinate 95%
APOSOR303727-1G
:
Apollo Scientific
:
Methyl-4,6-dichloro-5-nitronicotinate 95%
APOSOR303727-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
251.03 g/mol
:
65…66 °C
:
GHS07
:
Ambient
:
89247-05-2
:
C₇H₄Cl₂N₂O₄
:
MFCD13193110