Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-5-bromo-4,6-dihydroxynicotinate
Ethyl-5-bromo-4,6-dihydroxynicotinate
Ethyl-5-bromo-4,6-dihydroxynicotinate
Irritant
Ethyl-5-bromo-4,6-dihydroxynicotinate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Ethyl-5-bromo-4,6-dihydroxynicotinate
Supplier:
Apollo Scientific
About this item
CAS Number:
89978-58-5
:
Formula:
C₈H₈BrNO₄
MDL Number:
MFCD01411758
:
262.07 g/mol
Melting Pt:
267…269 °C
:
GHS07
Storage Temperature:
Ambient