Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-3-cyano-4,6-dimethyl-2-pyridinecarboxylate
Ethyl-3-cyano-4,6-dimethyl-2-pyridinecarboxylate
Ethyl-3-cyano-4,6-dimethyl-2-pyridinecarboxylate
Ethyl-3-cyano-4,6-dimethyl-2-pyridinecarboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
Ethyl-3-cyano-4,6-dimethyl-2-pyridinecarboxylate
Supplier:
Apollo Scientific
About this item
CAS Number:
1221792-05-7
:
Formula:
C₁₁H₁₂N₂O₂
MDL Number:
MFCD14584824
:
204.23 g/mol
Melting Pt:
58…60 °C
Storage Temperature:
Ambient