Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-nitrobenzonitrile
2-Chloro-4-nitrobenzonitrile
2-Chloro-4-nitrobenzonitrile
:
Apollo Scientific
2-Chloro-4-nitrobenzonitrile
:
Apollo Scientific
:
28163-00-0
:
:
C₇H₃ClN₂O₂
:
MFCD00052203
:
182.57 g/mol
:
78…82 °C
:
GHS07
:
Ambient