
4,5-Dimethyl-o-phenylenediamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3171-45-7
:
Formula:C₈H₁₂N₂
MDL Number:MFCD00007729
:136.2 g/mol
Melting Pt:127…129 °C
EINECS:221-635-1
:GHS07
Storage Temperature:Refrigerator