4-Amino-2,6-dimethylpyrimidine
Catalog # APOSOR302498-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:123.16 g/mol
Storage Temperature:Ambient
CAS Number:461-98-3
Formula:C₆H₉N₃
MDL Number:MFCD00006106