Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dibromo-2,6-dimethoxypyridine
3,5-Dibromo-2,6-dimethoxypyridine
3,5-Dibromo-2,6-dimethoxypyridine
3,5-Dibromo-2,6-dimethoxypyridine
Supplier:
Apollo Scientific
Certificates
Expand image
3,5-Dibromo-2,6-dimethoxypyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
16727-44-9
:
Formula:
C₇H₇Br₂NO₂
MDL Number:
MFCD00955954
:
296.95 g/mol
Storage Temperature:
Ambient