Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-4,6-dichloroquinoline-3-carboxylate
Ethyl-4,6-dichloroquinoline-3-carboxylate
Ethyl-4,6-dichloroquinoline-3-carboxylate
APOSOR302253-1G
:
Apollo Scientific
:
Ethyl-4,6-dichloroquinoline-3-carboxylate
APOSOR302253-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
270.109 g/mol
:
Ambient
:
21168-41-2
:
C₁₂H₉Cl₂NO₂
:
MFCD00173348