2,2-Dimethyl-N-pyridin-2-ylpropionamide
Catalog # APOSOR302212-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:178.235 g/mol
Storage Temperature:Ambient
CAS Number:86847-59-8
Formula:C₁₀H₁₄N₂O
MDL Number:MFCD00460277