2-Hydroxy-3-methylpyrazine
Catalog # APOSOR302157-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:110.11 g/mol
Storage Temperature:Ambient
CAS Number:19838-07-4
Formula:C₅H₆N₂O
MDL Number:MFCD01646120