Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Dibromonaphthalene

Supplier:   Apollo Scientific
2,3-Dibromonaphthalene
Supplier:  Apollo Scientific
Certificates
2,3-Dibromonaphthalene

6 Options Available Below

Product Details & Documents
Synonyms:  2,3-Diiodonaphthalene
  • CAS Number:13214-70-5
  • Formula:C₁₀H₆Br₂
  • MDL Number:MFCD00021589
  • Molecular Weight:285.97 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 21,80 €
    21,80 €
    1 * 100 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 24,60 €
    24,60 €
    1 * 250 mg
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 60,80 €
    60,80 €
    1 * 1 g
       60,80 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 109,00 €
    109,00 €
    1 * 5 g
       109,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 534,00 €
    534,00 €
    1 * 25 g
       534,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 2 130,00 €
    2 130,00 €
    1 * 100 g
       2 130,00 €

    Specifications:
    New Product
    New Product