1-Benzyl-4,5-dioxopyrrolidine-3-carboxylic acid ethyl ester
Catalog # APOSOR302032-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:261.28 g/mol
Storage Temperature:Ambient
CAS Number:5336-50-5
Formula:C₁₄H₁₅NO₄
MDL Number:MFCD00098219