Order Entry
Export
ContactUsLinkComponent
4-Iodo-o-xylene
Irritant
4-Iodo-o-xylene
Catalog # APOSOR30140-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Iodo-o-xylene
Catalog # APOSOR30140-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :232.06 g/mol
Boiling Pt:106…108 °C/13mmHg
Density:1.633 g/cm³ (25 °C)
Flash Pt:113 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:31599-61-8
Formula:C₈H₉I
MDL Number:MFCD00040989