Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dibromopyrazine
2,3-Dibromopyrazine 95%
2,3-Dibromopyrazine 95%
2,3-Dibromopyrazine 95%
APOSOR301382-1G
:
Apollo Scientific
:
2,3-Dibromopyrazine 95%
APOSOR301382-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
237.8800 g/mol
:
57…58 °C
:
GHS07
:
Ambient
:
95538-03-7
:
C₄H₂Br₂N₂
:
MFCD02684897