Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Oxa-7-azaspiro[3.5]nonane hydrochloride
1-Oxa-7-azaspiro[3.5]nonane hydrochloride
1-Oxa-7-azaspiro[3.5]nonane hydrochloride
Irritant
1-Oxa-7-azaspiro[3.5]nonane hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Oxa-7-azaspiro[3.5]nonane hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
38674-21-4
:
Formula:
C₇H₁₄ClNO
MDL Number:
MFCD28139317
:
163.65 g/mol
:
GHS07
Storage Temperature:
Ambient