Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Oxa-6-azaspiro[3.4]octane
2-Oxa-6-azaspiro[3.4]octane 95%
2-Oxa-6-azaspiro[3.4]octane 95%
APOSOR301342-250MG
:
Apollo Scientific
:
2-Oxa-6-azaspiro[3.4]octane 95%
APOSOR301342-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
113.16 g/mol
:
187 °C
:
1.08 g/cm³ (25 °C)
:
63 °C
:
GHS07
:
Ambient
:
220290-68-6
:
C₆H₁₁NO
:
MFCD14586455