Order Entry
Export
ContactUsLinkComponent
Pyrazolo[1,5-a]pyridine 97%
Pyrazolo[1,5-a]pyridine 97%
Catalog # APOSOR301332-1G
Supplier:  Apollo Scientific
CAS Number:  
Pyrazolo[1,5-a]pyridine 97%
Catalog # APOSOR301332-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :118.14 g/mol
Storage Temperature:Ambient
CAS Number:274-56-6
Formula:C₇H₆N₂
MDL Number:MFCD08752622