0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-Chloro-1H-pyrazolo[4,3-c]pyridine
4-Chloro-1H-pyrazolo[4,3-c]pyridine 98%
4-Chloro-1H-pyrazolo[4,3-c]pyridine 98%
:
Apollo Scientific
4-Chloro-1H-pyrazolo[4,3-c]pyridine 98%
:
Apollo Scientific
:
871836-51-0
:
:
C₆H₄ClN₃
:
MFCD11870058
:
153.5691 g/mol
:
GHS07
:
Ambient