Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-1H-pyrrolo[2,3-b]pyridine
2-Methyl-1H-pyrrolo[2,3-b]pyridine
2-Methyl-1H-pyrrolo[2,3-b]pyridine
Irritant
2-Methyl-1H-pyrrolo[2,3-b]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Methyl-1H-pyrrolo[2,3-b]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
23612-48-8
:
Formula:
C₈H₈N₂
MDL Number:
MFCD08669524
:
132.1600 g/mol
Melting Pt:
138…142 °C
:
GHS07
Storage Temperature:
Refrigerator