Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dichlorothiophenol

2,3-Dichlorothiophenol

Bottle, 5 g
Catalog # APOSOR30119-5G
Corrosive
Irritant
Supplier:   Apollo Scientific
2,3-Dichlorothiophenol
Catalog # APOSOR30119-5G
Supplier:  Apollo Scientific
CAS Number:  17231-95-7
2,3-Dichlorothiophenol

Bottle, 5 g

CAS Number:  17231-95-7
Price
43,50 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,3-Dichlorobenzenethiol, 1,2-Dichloro-3-mercaptobenzene, 2,3-Dichlorobenzene-1-thiol
  • CAS Number:17231-95-7
  • Formula:C₆H₄Cl₂S
  • MDL Number:MFCD00041420
  • Molecular Weight:179.07 g/mol
  • Boiling Pt:96…98 °C/0.5mmHg
  • Melting Pt:54…60 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications