Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-6-methoxypyrimidine
2,4-Dichloro-6-methoxypyrimidine
2,4-Dichloro-6-methoxypyrimidine
2,4-Dichloro-6-methoxypyrimidine
APOSOR301155-250MG
:
Apollo Scientific
:
2,4-Dichloro-6-methoxypyrimidine
APOSOR301155-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
179.0000 g/mol
:
GHS07
:
Refrigerator
:
43212-41-5
:
C₅H₄Cl₂N₂O
:
MFCD00955635