Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromo-1H-pyrrolo[3,2-c]pyridine
7-Bromo-1H-pyrrolo[3,2-c]pyridine
7-Bromo-1H-pyrrolo[3,2-c]pyridine
APOSOR301138-1G
:
Apollo Scientific
:
7-Bromo-1H-pyrrolo[3,2-c]pyridine
APOSOR301138-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
197.0320 g/mol
:
Ambient
:
902837-42-7
:
C₇H₅BrN₂
:
MFCD07781153