Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
Pyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
Pyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
Pyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
APOSOR301058-1G
:
Apollo Scientific
:
Pyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
APOSOR301058-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
135.13 g/mol
:
Ambient
:
159326-71-3
:
C₆H₅N₃O
:
MFCD12923122