Order Entry
Export
Orders LinkContactUsLinkComponent
6-Methoxyoxindole

6-Methoxyoxindole

Irritant
Supplier:   Apollo Scientific
6-Methoxyoxindole
Supplier:  Apollo Scientific
Certificates
6-Methoxyoxindole

4 Options Available Below

Product Details & Documents
  • CAS Number:7699-19-6
  • Formula:C₉H₉NO₂
  • MDL Number:MFCD00239414
  • Molecular Weight:163.18 g/mol
  • Melting Pt:160…161 °C
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 138,00 €
    138,00 €
    1 * 250 mg
       138,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 828,00 €
    828,00 €
    1 * 5 g
       828,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1 590,00 €
    1 590,00 €
    1 * 10 g
       1 590,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 2 790,00 €
    2 790,00 €
    1 * 25 g
       2 790,00 €