0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Methyl m-toluate
Methyl-m-toluate
Methyl-m-toluate
APOSOR30098-5G
:
Apollo Scientific
:
Methyl-m-toluate
APOSOR30098-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
113 °C/27mmHg
:
1.062 g/cm³ (25 °C)
:
95 °C
:
150.18
:
GHS07
:
Ambient
:
99-36-5
:
C₉H₁₀O₂
:
MFCD00008436