
1,3-Dichloro-4-iodobenzene
Catalog # APOSOR30094-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:272.9 g/mol
Boiling Pt:262…263 °C
Density:2.01 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:29898-32-6
Formula:C₆H₃Cl₂I
MDL Number:MFCD00001035