0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Supplier:
Apollo Scientific
About this item
CAS Number:
59857-86-2
:
Formula:
C₇H₁₁NO₃
MDL Number:
MFCD00015898
:
157.169 g/mol
Storage Temperature:
Ambient