Order Entry
Export
ContactUsLinkComponent
Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate 95%
Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate 95%
Catalog # APOSOR300724-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate 95%
Catalog # APOSOR300724-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :208.2100 g/mol
Storage Temperature:Ambient
CAS Number:4739-94-0
Formula:C₁₁H₁₂O₄
MDL Number:MFCD01099774