Order Entry
Export
Orders LinkContactUsLinkComponent
5-(3-Bromophenyl)cyclohexane-1,3-dione

5-(3-Bromophenyl)cyclohexane-1,3-dione

Supplier:   Apollo Scientific
5-(3-Bromophenyl)cyclohexane-1,3-dione
Supplier:  Apollo Scientific
Certificates

4 Options Available Below

Product Details & Documents
  • CAS Number:144128-71-2
  • Formula:C₁₂H₁₁BrO₂
  • MDL Number:MFCD03844399
  • Molecular Weight:267.13 g/mol
  • Storage Temperature:Ambient
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 94,10 €
    94,10 €
    1 * 250 mg
       94,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR300642-250MG
  • Bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 142,00 €
    142,00 €
    1 * 500 mg
       142,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR300642-500MG
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 189,00 €
    189,00 €
    1 * 1 g
       189,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR300642-1G
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 763,00 €
    763,00 €
    1 * 5 g
       763,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR300642-5G