Order Entry
Export
ContactUsLinkComponent
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid 97%
Irritant
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid 97%
Catalog # APOSOR300511-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid 97%
Catalog # APOSOR300511-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :241.05 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:944896-42-8
Formula:C₈H₅BrN₂O₂
MDL Number:MFCD10698063