7-Aminopyrazolo[1,5-a]pyrimidine-6-carbonitrile
Catalog # APOSOR300510-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:159.15 g/mol
Storage Temperature:Ambient
CAS Number:89975-57-5
Formula:C₇H₅N₅
MDL Number:MFCD00104599