1,6-Dichloroisoquinoline 97%
Catalog # APOSOR300125-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:198.0500 g/mol
Storage Temperature:Ambient
CAS Number:630421-73-7
Formula:C₉H₅Cl₂N
MDL Number:MFCD06738660