Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dihydroisoquinolin-1(2H)-one
3,4-Dihydroisoquinolin-1(2H)-one
3,4-Dihydroisoquinolin-1(2H)-one
Irritant
3,4-Dihydroisoquinolin-1(2H)-one
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,4-Dihydroisoquinolin-1(2H)-one
Supplier:
Apollo Scientific
About this item
CAS Number:
1196-38-9
:
Formula:
C9H9NO
MDL Number:
MFCD00853963
Melting Pt:
75…80 °C
:
147.1700 g/mol
:
GHS07
Storage Temperature:
Refrigerator