6-Bromo-8-methoxyimidazo[1,2-a]pyrazine
Catalog # APOSOR300022-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:228.0500 g/mol
Storage Temperature:Ambient
CAS Number:63744-25-2
Formula:C₇H₆BrN₃O
MDL Number:MFCD09835036