3-Bromo-8-chloroimidazo[1,2-a]pyrazine
Catalog # APOSOR300016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.4651 g/mol
Storage Temperature:Ambient
CAS Number:143591-61-1
Formula:C6H3BrClN3
MDL Number:MFCD09909653