Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dihydroxypyridine
2,4-Dihydroxypyridine 98%
2,4-Dihydroxypyridine 98%
Irritant
2,4-Dihydroxypyridine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Dihydroxypyridine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
626-03-9
:
84719-31-3
Formula:
C₅H₅NO₂
MDL Number:
MFCD00006273
:
111.10 g/mol
Melting Pt:
272…276 °C
EINECS:
210-924-8
:
GHS07
Storage Temperature:
Ambient