Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloro-2-(methylthio)pyrimidine
4,6-Dichloro-2-(methylthio)pyrimidine
4,6-Dichloro-2-(methylthio)pyrimidine
:
Apollo Scientific
4,6-Dichloro-2-(methylthio)pyrimidine
:
Apollo Scientific
:
6299-25-8
:
:
C₅H₄Cl₂N₂S
:
MFCD00006086
:
195.07 g/mol
:
135…136 °C/14mmHg
:
38…42 °C
:
>110 °C
:
Ambient
:
228-577-6
:
3263
:
8,II
:
GHS05
:
GHS07