3,4-Dimethoxybenzonitrile
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Veratronitrile
- CAS Number:2024-83-1
- Formula:C₉H₉NO₂
- MDL Number:MFCD00001802
- Molecular Weight:163.18 g/mol
- Melting Pt:65…70 °C
- Storage Temperature:Ambient
- EINECS:217-969-2
- UN:3439
- ADR:6.1,III
5 Options Available Below
Product Details & Documents
Synonyms: Veratronitrile
- CAS Number:2024-83-1
- Formula:C₉H₉NO₂
- MDL Number:MFCD00001802
- Molecular Weight:163.18 g/mol
- Melting Pt:65…70 °C
- Storage Temperature:Ambient
- EINECS:217-969-2
- UN:3439
- ADR:6.1,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



