Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,5-Dichloronicotinamide

Irritant
Supplier:   Apollo Scientific
2,5-Dichloronicotinamide
Supplier:  Apollo Scientific
Certificates
2,5-Dichloronicotinamide

2 Options Available Below

Product Details & Documents
  • CAS Number:75291-86-0
  • Formula:C₆H₄Cl₂N₂O
  • MDL Number:MFCD00204159
  • Melting Pt:171…173 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 57,90 €
    57,90 €
    1 * 1 g
       57,90 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 232,00 €
    232,00 €
    1 * 5 g
       232,00 €